Merck
  • Headspace solid-phase microextraction-gas chromatography-mass spectrometry characterization of propolis volatile compounds.
Headspace solid-phase microextraction-gas chromatography-mass spectrometry characterization of propolis volatile compounds.
Journal of pharmaceutical and biomedical analysis (2013-06-29)
Federica Pellati, Francesco Pio Prencipe, Stefania Benvenuti
ABSTRACT
In this study, a novel and efficient method based on headspace solid-phase microextraction (HS-SPME), followed by gas chromatography-mass spectrometry (GC-MS), was developed for the analysis of propolis volatile compounds. The HS-SPME procedure, whose experimental parameters were properly optimized, was carried out using a 100 μm polydimethylsiloxane (PDMS) fiber. The GC-MS analyses were performed on a HP-5 MS cross-linked 5% diphenyl-95% dimethyl polysiloxane capillary column (30 m × 0.25 mm I.D., 1.00 μm film thickness), under programmed-temperature elution. Ninety-nine constituents were identified using this technique in the samples of raw propolis collected from different Italian regions. The main compounds detected include benzoic acid (0.87-30.13%) and its esters, such as benzyl benzoate (0.16-13.05%), benzyl salicylate (0.34-1.90%) and benzyl cinnamate (0.34-3.20%). Vanillin was detected in most of the samples analyzed in this study (0.07-5.44%). Another relevant class of volatile constituents is represented by sesquiterpene hydrocarbons, such as δ-cadinene (1.29-13.31%), γ-cadinene (1.36-8.85%) and α-muurolene (0.78-6.59%), and oxygenated sesquiterpenes, such as β-eudesmol (2.33-12.83%), T-cadinol (2.73-9.95%) and α-cadinol (4.84-9.74%). Regarding monoterpene hydrocarbons, they were found to be present at low level in the samples analyzed in this study, with the exception of one sample from Southern Italy, where α-pinene was the most abundant constituent (13.19%). The results obtained by HS-SPME-GC-MS were also compared with those of hydrodistillation (HD) coupled with GC-MS. The HS-SPME-GC-MS method developed in this study allowed us to determine the chemical fingerprint of propolis volatile constituents, thus providing a new and reliable tool for the complete characterization of this biologically active apiary product.
Product Description
Supelco
Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 23 ga
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with manual holder, needle size 23 ga
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
SPME Portable Field Sampler, coating PDMS
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm(PDMS, needle size 23 ga, for use with autosampler
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Supelco
SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm(PDMS, for use with autosampler, needle size 24 ga
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® 1000 Reversible SPE Tube, 200 mg/mL, pk 30
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of 1 ×, plain gray hub