Merck
  • Lipidemic responses of male broiler chickens to enzyme-supplemented wheat-soybean meal-based diets with various levels of metabolizable energy.
Lipidemic responses of male broiler chickens to enzyme-supplemented wheat-soybean meal-based diets with various levels of metabolizable energy.
Pakistan journal of biological sciences : PJBS (2014-02-12)
K Karimi, B Shokrollahi
ABSTRACT
Effects of 2 various levels of AME (according to the manual recommendation and 100 kcal kg(-1) less than it), 2 levels of endo-beta-D-mannanase enzyme (0, 1 g kg(-1)) and 2 levels of xylanase enzyme (0 and 1 g kg(-1)) on serum lipid parameters as a 2(3) factorial arrangement were tested in 120 male broiler chicks fed wheat-soybean meal-based diet. These birds were randomly assigned to 8 experimental groups with 3 pen per group and 5 birds per pen. The serum HDL-cholesterol (HDL), LDL-cholesterol (LDL), Total-cholesterol (TC) and Triglycerides (TG) concentrations were measured at 31 and 41 day of age. The concentrations of serum TG, TC and LDL of 41-day-old birds demonstrated to be lower than those of 31-d-old (p < 0.001). Some hypolipidemic responses were observed in the broiler chicks fed on (1) Diet supplemented with only beta-mannanase, (2) Normal-AME diets supplemented with p-mannanase, (3) Normal-AME diets supplemented with Xylanse and (4) Normal-AME diets supplemented with both beta-mannanase and Xylanase (p < 0.01). In the other hand, some hyperlipidemic responses were detected in the broiler chicks fed on low-AME diets supplemented with xylanse or beta-mannanase enzymes, alone or in combination (p < 0.01). Regardless of AME, adding both xylanse and beta-mannanase to the wheat-soybean meal-based diets have both hyperlipidemic and hypolipidemic effects together (p < 0.01).
Sigma-Aldrich
Lipase from Mucor miehei, powder, slightly brown, ~1 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized, powder (fine), 15-25 U/mg
Sigma-Aldrich
Lipase from Rhizopus oryzae, powder (fine), ~10 U/mg
Sigma-Aldrich
Lipase from Candida rugosa, powder, yellow-brown, ≥2 U/mg
Sigma-Aldrich
Lipase from Aspergillus oryzae, lyophilized, powder, white, ~50 U/mg
Sigma-Aldrich
Lipase from Mucor miehei, lyophilized powder, ≥4,000 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from porcine pancreas, Type II, ≥125 units/mg protein (using olive oil (30 min incubation)), 30-90 units/mg protein (using triacetin)
Sigma-Aldrich
Lipase from porcine pancreas, Type VI-S, ≥20,000 units/mg protein, lyophilized powder
Sigma-Aldrich
Lipase from wheat germ, Type I, lyophilized powder, 5-15 units/mg solid
Sigma-Aldrich
Lipase acrylic resin, ≥5,000 U/g, recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase from Aspergillus oryzae, solution, ≥100,000 U/g, white, beige
Sigma-Aldrich
Lipase from Aspergillus oryzae, ≥20,000 U/g
Sigma-Aldrich
Lipase from Pseudomonas sp., Type XIII, lyophilized powder, ≥15 units/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, Type VII, ≥700 unit/mg solid
Sigma-Aldrich
Lipase from Candida rugosa, lyophilized powder, ≥40,000 units/mg protein
Sigma-Aldrich
Lipase from Rhizopus niveus, powder (fine), ≥1.5 U/mg
Sigma-Aldrich
Lipase from Candida sp., recombinant, expressed in Aspergillus niger
Sigma-Aldrich
Lipase B Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~9 U/mg
Sigma-Aldrich
Lipase A Candida antarctica, recombinant from Aspergillus oryzae, powder, beige, ~2 U/mg
Sigma-Aldrich
Lipase from Pseudomonas cepacia, powder, light beige, ≥30 U/mg
Sigma-Aldrich
β-Mannosidase from Helix pomatia, 5-30 units/mL, ammonium sulfate suspension, crude extract
Sigma-Aldrich
Lipase from Mucor javanicus, lyophilized powder, ≥300 units/mg solid (using olive oil)
Sigma-Aldrich
Lipase from Aspergillus niger, powder (fine), ~200 U/g
Sigma-Aldrich
Lipase immobilized from Candida antarctica, beads, slightly brown, >2 U/mg