Merck
  • Interleukin-6 receptor blockade enhances CD39+ regulatory T cell development in rheumatoid arthritis and in experimental arthritis.
Interleukin-6 receptor blockade enhances CD39+ regulatory T cell development in rheumatoid arthritis and in experimental arthritis.
Arthritis & rheumatology (Hoboken, N.J.) (2014-02-08)
A Thiolat, L Semerano, Y M Pers, J Biton, D Lemeiter, P Portales, J Quentin, C Jorgensen, P Decker, M-C Boissier, P Louis-Plence, N Bessis
ABSTRACT
The rationale for blocking interleukin-6 (IL-6) in rheumatoid arthritis (RA) lies chiefly in the proinflammatory effect of this cytokine. Few studies have evaluated the consequences of anti-IL-6 receptor (IL-6R) antibody treatment on Treg cells. This study was undertaken to elucidate the mechanism of action of anti-IL-6R antibody treatment by studying the effects on Treg cells in an experimental arthritis model and in patients with RA. Mice with collagen-induced arthritis (CIA) were treated with a mouse anti-IL-6R antibody (MR16-1), and changes in Treg, Th1, and Th17 cells were assessed at key time points during the course of the disease. Peripheral blood from 15 RA patients was collected on day 0 and after 3 months of tocilizumab treatment for flow cytometry analysis of Th17 and Treg cells. In MR16-1-treated mice, Th17 cell frequencies were unchanged, whereas Treg cell frequencies were increased. The Treg cell phenotype showed marked changes, with an increase in the frequency of CD39+ Treg cells in the lymph nodes and spleen. Interestingly, similar CD39+ Treg cell expansion was observed in RA patients who were tocilizumab responders at 3 months, with no change in Th17 cell frequency. Moreover, fluorescence-activated cell-sorted CD39+ Treg cells from responder RA patients were functionally able to suppress the proliferation of conventional T cells. In both CIA and RA, the frequency of functionally suppressive CD39+ Treg cells is increased as a result of anti-IL-6R treatment, whereas Th17 cells are unaffected. The modification of Treg cell frequency and phenotype may be one of the mechanisms involved in the therapeutic effect of IL-6 blockade in RA.
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, suitable for protein labeling, ≥90% (HPLC), powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥200 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥3.0 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potatoes, ATPase ≥60 units/mg protein, lyophilized powder (partially purified)
Sigma-Aldrich
Apyrase from potatoes, High Activity, ATPase ≥600 units/mg protein, lyophilized powder
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)