Merck
  • Salivary nerve growth factor reactivity to acute psychosocial stress: a new frontier for stress research.
Salivary nerve growth factor reactivity to acute psychosocial stress: a new frontier for stress research.
Psychosomatic medicine (2013-10-01)
Heidemarie K Laurent, Sean M Laurent, Douglas A Granger
ABSTRACT
Neurotrophins such as nerve growth factor (NGF) may represent a stress-responsive system complementing the better known neuroendocrine (hypothalamic-pituitary-adrenal axis) and autonomic nervous system, but there is little evidence for NGF response to acute stress in humans because noninvasive measures have not been available. We investigated salivary NGF (sNGF) in 40 healthy young adults confronting a romantic conflict stressor. Five saliva samples-two collected before and three after the conflict-were assayed for sNGF, cortisol (hypothalamic-pituitary-adrenal marker), and α-amylase (sAA; ANS marker). In addition, a control group (n = 20) gave saliva samples at the same time intervals to determine whether sNGF changes were specific to the conflict stressor. sNGF showed significant reactivity from entry to the first poststress sample among study participants (β = .13, p = .001), with nonsignificant change across poststress samples. Control participants showed no change in sNGF across the same period. Within-person changes in sNGF were generally aligned with both cortisol (β = .17, p = .003) and sAA (β = .17, p = .021) responses. Preconflict negative emotion predicted lower sNGF reactivity (β = -.08, p = .009) and less alignment with sAA (β = -.09, p = .040), whereas positive emotion predicted less alignment with cortisol (β = -.10, p = .019). This study is the first to document sNGF as a marker that responds to stress in humans.
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, aqueous solution, ≥800 FAU/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Bacillus amyloliquefaciens, liquid, ≥250 units/g
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, ≥150 units/mg protein (biuret)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~30 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from human saliva, Type XIII-A, lyophilized powder, 300-1,500 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, lyophilized powder, 500-1,500 units/mg protein, 93-100% (SDS-PAGE)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from human saliva, Type IX-A, lyophilized powder, 1,000-3,000 units/mg protein
SKU
Pack Size
Availability
Price
Quantity
Supelco
α-Amylase from Bacillus licheniformis, suitable for determination of starch (Kit STA-20)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Bacillus licheniformis, Type XII-A, saline solution, ≥500 units/mg protein (biuret)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Aspergillus oryzae, powder, ~1.5 U/mg (~0.2 U acc. to Willstätter)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Taka-Diastase from Aspergillus oryzae, powder, slightly beige, ~100 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Bacillus sp., Type II-A, lyophilized powder, ≥1,500 units/mg protein (biuret)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from porcine pancreas, PMSF Treated, Type I-A, saline suspension, ≥1000 units/mg protein (E1%/280)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from porcine pancreas, Type VI-B, ≥5 units/mg solid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from porcine pancreas, Type I-A, PMSF treated, saline suspension, 700-1400 units/mg protein (E1%/280)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, ≥400 units/mg protein (Lowry)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~380 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
α-Amylase from Bacillus sp., liquid