Merck
  • Polymorphisms in alcohol-metabolizing enzymes and esophageal carcinoma susceptibility: a Dutch Caucasian case-control study.
Polymorphisms in alcohol-metabolizing enzymes and esophageal carcinoma susceptibility: a Dutch Caucasian case-control study.
Journal of human genetics (2013-09-21)
Polat Dura, Tineke Berkers, Elke M van Veen, Jody Salomon, Rene H M te Morsche, Hennie M J Roelofs, Jon O Kristinsson, Theo Wobbes, Ben J M Witteman, Adriaan C I T L Tan, Joost P H Drenth, Wilbert H M Peters
ABSTRACT
Esophageal cancer (EC), mainly consisting of squamous cell carcinoma (ESCC) in the Eastern world and adenocarcinoma (EAC) in the Western world, is strongly associated with dietary factors such as alcohol use. We aimed to clarify the modifying role in EC etiology in Caucasians of functional genotypes in alcohol-metabolizing enzymes. In all, 351 Caucasian patients with EC and 430 matched controls were included and polymorphisms in CYP2E1, ADH and near ALDH2 genes were determined. In contrast to the results on ESCC in mainly Asian studies, we found that functional genotypes of alcohol-metabolizing enzymes were not significantly associated with EAC or ESCC in an European population.
Sigma-Aldrich
Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Alcohol Dehydrogenase from Saccharomyces cerevisiae