Merck
  • Use of steroids to monitor alterations in the outer membrane of Pseudomonas aeruginosa.
Use of steroids to monitor alterations in the outer membrane of Pseudomonas aeruginosa.
Journal of bacteriology (1997-11-26)
P Plesiat, J R Aires, C Godard, T Köhler
ABSTRACT
Testosterone (a strongly hydrophobic steroid) and testosterone hemisuccinate (a negatively charged derivative) were used as probes to investigate alterations in the outer membrane of Pseudomonas aeruginosa. Diffusion rates of the steroids across the lipid bilayer were measured by coupling the influx of these compounds to their subsequent oxidation by an intracellular delta1-dehydrogenase enzyme. Wild-type cells of P. aeruginosa (strain PAO1) were found to be 25 times more permeable to testosterone than to testosterone hemisuccinate. The uptake of the latter compound appeared to be partially dependent on the external pH, thus suggesting a preferential diffusion of the uncharged protonated form across the cell envelope. Using various PAO mutants, we showed that the permeation of steroids was not affected by overexpression of active efflux systems but was increased up to 5.5-fold when the outer membrane contained defective lipopolysaccharides or lacked the major porin OprF. Such alterations in the hydrophobic uptake pathway were not, however, associated with an enhanced permeability of the mutants to the small hydrophilic molecule N,N,N',N'-tetramethyl-p-phenylene diamine. Thirty-six agents were also assayed for their ability to damage the cell surface of strain PAO1, using testosterone as a probe. Polymyxins, rBPI23, chlorhexidine, and dibromopropamidine demonstrated the strongest permeabilizing activities on a molar basis in the presence of 1 mM MgCl2. These amphiphilic polycations increased the transmembrane diffusion of testosterone up to 50-fold and sensitized the PAO1 cells to hydrophobic antibiotics. All together, these data indicated that the steroid uptake assay provides a direct and accurate measurement of the hydrophobic uptake pathway in P. aeruginosa.
Product Description
Dibrompropamidine diisetionate, European Pharmacopoeia (EP) Reference Standard
Dibrompropamidine for system suitability, European Pharmacopoeia (EP) Reference Standard