Merck
  • Enantioselective accumulation of (--)-pinoresinol through O-demethylation of (+/-)-eudesmin by Aspergillus niger.
Enantioselective accumulation of (--)-pinoresinol through O-demethylation of (+/-)-eudesmin by Aspergillus niger.
Phytochemistry (1997-04-01)
H Kasahara, M Miyazawa, H Kameoka
ABSTRACT
Microbial transformation of (+/-)-eudesmin by Aspergillus niger was investigated. Enantioselective accumulation of (--)-pinoresinol was shown through O-demethylation of (+/-)-eudesmin. This fungus O- demethylated both enantiomers of eudesmin, but the conversion rates for each enantiomer were clearly different.
Sigma-Aldrich
Eudesmine, ≥95% (LC/MS-ELSD)