Merck
  • Positive influence of AT(1) receptor antagonism upon the impaired celiprolol-induced vasodilatation in aorta from spontaneously hypertensive rats.
Positive influence of AT(1) receptor antagonism upon the impaired celiprolol-induced vasodilatation in aorta from spontaneously hypertensive rats.
European journal of pharmacology (2010-07-20)
Frédérique Sauvaget, Mohamed Yassine Mallem, Véronique Bucas, Marc Gogny, Jean-Claude Desfontis, Jacques Noireaud
ABSTRACT
We evaluated celiprolol-induced vasodilatation in aorta taken from 12-week-old spontaneously hypertensive rats (SHR) and the effect of AT(1) angiotensin II receptor antagonism on the vasodilatory action of celiprolol in Wistar Kyoto (WKY) rats and SHR. In WKY rats, the celiprolol-induced relaxation was greatly decreased in denuded aorta, and completely abolished in intact aorta by N(omega)-nitro-l-arginine methyl ester (l-NAME, 100 microM). In SHR, celiprolol-induced relaxation was reduced compared to WKY rats (E(max) (value obtained for the highest concentration, 300 microM)=39.1+ or - 3.78%, n=21 vs. 80.4 + or - 3% in WKY rats, n=10; P<0.0001). Endothelium removal or pre-treatment with l-NAME did not alter celiprolol-induced relaxation in SHR. In both strains, relaxation to celiprolol was decreased in the presence of nadolol (a beta(1)/beta(2)-adrenoceptor antagonist, 10 microM). N-[[3-[(2S)-2-hydroxy-3-[[2-[4-[(phenylsulfonyl)amino] phenyl]ethyl]amino] propoxy]phenyl]methyl]-acetamide (L748337, a beta(3)-adrenoceptor antagonist, 7 microM) had no effect. A 12-day treatment with candesartan cilexetil (an AT(1) receptor antagonist, 0.37 or 1mg/kg/day) reduced systolic blood pressure in both strains, but only improved relaxation to celiprolol in SHR, and only at the highest dose (E(max)=64.2+/-3.9%, n=10, P<0.0001 vs. SHR control). In both strains, local aortic AT(1) receptor antagonism with candesartan CV11974 (100 microM) had no effect. The endothelial beta(1)/beta(2) relaxation induced by celiprolol was therefore impaired in SHR aorta and AT(1) receptor antagonism improved the response to celiprolol, in conjunction with a reduction in blood pressure. This work highlights the need to analyse the potential benefit of a combination of celiprolol/AT(1) receptor antagonist in the treatment of hypertension.