Merck
  • Eudesmin inhibits tumor necrosis factor-alpha production and T cell proliferation.
Eudesmin inhibits tumor necrosis factor-alpha production and T cell proliferation.
Archives of pharmacal research (1999-09-18)
J Y Cho, E S Yoo, K U Baik, M H Park
ABSTRACT
Possible antiinflammatory effects of eudesmin were examined by assessing the effects on tumor necrosis factor (TNF)-alpha production and lymphocyte proliferation as well as cytotoxicity against murine and human macrophages. The compound significantly inhibited TNF-alpha production by lipopolysaccharide (LPS)-stimulated murine macrophage RAW264.7 without displaying cytotoxicity suggesting that eudesmin may inhibit TNF-alpha production without any interference of normal cell function. It also significantly attenuated T cell proliferation stimulated by concanavalin A (Con A) in a dose-dependent manner.
Sigma-Aldrich
Eudesmine, ≥95% (LC/MS-ELSD)