Merck
  • Analysing fluorobenzoate tracers in groundwater samples using liquid chromatography-tandem mass spectrometry. A tool for leaching studies and hydrology.
Analysing fluorobenzoate tracers in groundwater samples using liquid chromatography-tandem mass spectrometry. A tool for leaching studies and hydrology.
Journal of chromatography. A (2002-07-10)
René K Juhler, Annette P Mortensen
ABSTRACT
A sensitive LC-MS-MS method for the direct determination and quantification of 15 fluorobenzoic acids (FBAs) was developed. FBAs are used as conservative tracers for hydrological modelling of water flow and in studies of pesticides and other xenobiotic compounds. The use of FBAs is discussed in relation to other tracers (bromide, chloride, uranine). The method covers mono-substituted fluorobenzoic acid, difluorobenzoic acid, trifluorobenzoic acid, and tetrafluorobenzoic acid. The general detection limit in ground water was 1 microg/l using electrospray ionisation and 20 microg/l using atmospheric pressure chemical ionisation. Analysis time was less than 10 min, small sample volumes were needed and no clean-up was required.