Merck
  • TRIM16 acts as an E3 ubiquitin ligase and can heterodimerize with other TRIM family members.
TRIM16 acts as an E3 ubiquitin ligase and can heterodimerize with other TRIM family members.
PloS one (2012-05-26)
Jessica L Bell, Alena Malyukova, Jessica K Holien, Jessica Koach, Michael W Parker, Maria Kavallaris, Glenn M Marshall, Belamy B Cheung
ABSTRACT
The TRIM family of proteins is distinguished by its tripartite motif (TRIM). Typically, TRIM proteins contain a RING finger domain, one or two B-box domains, a coiled-coil domain and the more variable C-terminal domains. TRIM16 does not have a RING domain but does harbour two B-box domains. Here we showed that TRIM16 homodimerized through its coiled-coil domain and heterodimerized with other TRIM family members; TRIM24, Promyelocytic leukaemia (PML) protein and Midline-1 (MID1). Although, TRIM16 has no classic RING domain, three-dimensional modelling of TRIM16 suggested that its B-box domains adopts RING-like folds leading to the hypothesis that TRIM16 acts as an ubiquitin ligase. Consistent with this hypothesis, we demonstrated that TRIM16, devoid of a classical RING domain had auto-polyubiquitination activity and acted as an E3 ubiquitin ligase in vivo and in vitro assays. Thus via its unique structure, TRIM16 possesses both heterodimerization function with other TRIM proteins and also has E3 ubiquitin ligase activity.
Sigma-Aldrich
Monoclonal Anti-TRIM16 antibody produced in mouse, clone 5G11, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-TRIM16 antibody produced in mouse, clone 5F4, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-TRIM16 antibody produced in rabbit, IgG fraction of antiserum
Sigma-Aldrich
Anti-TRIM16 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution