Merck
  • Role of PKC-ζ in NADPH oxidase-PKCα-Giα axis dependent inhibition of β-adrenergic response by U46619 in pulmonary artery smooth muscle cells.
Role of PKC-ζ in NADPH oxidase-PKCα-Giα axis dependent inhibition of β-adrenergic response by U46619 in pulmonary artery smooth muscle cells.
Archives of biochemistry and biophysics (2013-11-05)
Sajal Chakraborti, Soumitra Roy, Amritlal Mandal, Animesh Chowdhury, Tapati Chakraborti
ABSTRACT
Treatment of bovine pulmonary artery smooth muscle cells (BPASMCs) with U46619 attenuated isoproterenol caused stimulation of adenyl cyclase activity and cAMP production. Pretreatment with SQ29548 (Tp receptor antagonist), apocynin (NADPH oxidase inhibitor) and Go6976 (PKC-α inhibitor) eliminated U46619 caused attenuation of isoproterenol stimulated adenyl cyclase activity. Pretreatment with SQ29548 and apocynin prevented U46619 induced increase in NADPH oxidase activity, PKC-α activity and Giα phosphorylation. However, pretreatment with CZI, a PKC-ζ inhibitor, markedly, but not completely, inhibited U46619 induced increase in NADPH oxidase activity, PKC-α activity, Giα phosphorylation and also significantly eliminated U46619 caused attenuation of isoproterenol stimulated adenyl cyclase activity. Pretreatment with Go6976 inhibited U46619 induced increase in Giα phosphorylation, but not PKC-ζ activity and NADPH oxidase activity. Pretreatment with pertussis toxin eliminated U46619 caused attenuation of isoproterenol stimulated adenyl cyclase activity without any discernible change in PKC-ζ, NADPH oxidase and PKC-α activities. Transfection of the cells with Tp, PKC-ζ and PKC-α siRNA duplexes corroborate the findings observed with their respective pharmacological inhibitors on the responses produced by U46619. Taken together, we suggest involvement of PKC-ζ in U46619 caused attenuation of isoproterenol stimulated β-adrenergic response, which is regulated by NADPH oxidase-PKCα-Giα axis in pulmonary artery smooth muscle cells.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
9,11-Dideoxy-11α,9α-epoxymethanoprostaglandin F, solution, 10 mg/mL in methyl acetate
Sigma-Aldrich
Isoprenaline hydrochloride
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Isoprenaline sulfate, European Pharmacopoeia (EP) Reference Standard