Merck
  • Trypsin functionalization and zirconia coating of mesoporous silica nanotubes for matrix-assisted laser desorption/ionization mass spectrometry analysis of phosphoprotein.
Trypsin functionalization and zirconia coating of mesoporous silica nanotubes for matrix-assisted laser desorption/ionization mass spectrometry analysis of phosphoprotein.
Journal of chromatography. A (2013-08-08)
Xiaoli Zhang, Fei Wang, Yan Xia
ABSTRACT
Trypsin functionalized mesoporous silica nanotubes bioreactor (TEMSN) and zirconia layer coated mesoporous silica nanotubes (ZrO2-MSN) were developed to deal with the long in-solution digestion time of phosphoprotein and detection difficulty of phosphorylated peptides, respectively. Trypsin was immobilized on the mesoporous silica nanotubes via epoxy group and TEMSN were used as a bioreactor for digestion of α-casein within 3min. ZrO2-MSN were performed to enrich phosphopeptides selectively from in-solution digested peptide mixture of β-casein to demonstrate that ZrO2-MSN possessed remarkable selectivity for phosphorylated peptides even at 100/1 molar ratio of BSA/β-casein. The selective ability of ZrO2-MSN was also investigated in comparison to ZrO2 nanoparticles (ZrO2 NP). Moreover, phosphorylated peptides at the femtomole (2.5fmol) level can also be detected with high S/N (signal-to-noise) ratio. Phosphopeptides enriched from TEMSN-bioreactor digested peptide mixture of α-casein was also performed to evaluate the cooperative performance of TEMSN and ZrO2-MSN platform. The experimental results indicated that TEMSN-bioreactor digestion changed the distribution of relative abundance of phosphopeptides and improved the relative intensity of partial phosphopeptides. This analytical strategy has also been applied to the identification of phosphopeptides isolated from non-fat bovine milk and got a comparable results compared with other materials cited from the literature. By matrix-assisted laser desorption/ionization mass spectrometry (MALDI-TOF MS), TEMSN and ZrO2-MSN were combined together for the rapid and comprehensive analysis of phosphoprotein.
Sigma-Aldrich
Zirconium, sponge, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, foil, thickness 0.1 mm, 99.98% trace metals basis
Sigma-Aldrich
Zirconium, rod, diam. 6.35 mm, ≥99% trace metals basis
Sigma-Aldrich
Zirconium, powder, −100 mesh
Sigma-Aldrich
Zirconium(IV) oxide, 99.99% trace metals basis (purity excludes ~2% HfO2)
Sigma-Aldrich
Zirconium(IV) oxide, powder, 5 μm, 99% trace metals basis
Sigma-Aldrich
Zirconium(IV) oxide, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 5 wt. % in H2O
Sigma-Aldrich
Zirconium(IV) oxide, nanoparticles, dispersion, <100 nm particle size (BET), 10 wt. % in H2O