Merck
  • Arylalkylidene rhodanine with bulky and hydrophobic functional group as selective HCV NS3 protease inhibitor.
Arylalkylidene rhodanine with bulky and hydrophobic functional group as selective HCV NS3 protease inhibitor.
Bioorganic & medicinal chemistry letters (2001-02-24)
W T Sing, C L Lee, S L Yeo, S P Lim, M M Sim
ABSTRACT
Arylalkylidene rhodanines 2(a-d) inhibit HCV NS3 protease at moderate concentrations. They are better inhibitors of other serine proteases such as chymotrypsin and plasmin. However, the selectivity of arylmethyliden e rhodanines (8a, 9a) with bulkier and more hydrophobic functional groups increases by 13- and 25-fold towards HCV NS3 protease respectively.
Sigma-Aldrich
Rhodanine, 97%
Sigma-Aldrich
Methyl thioglycolate, 95%