Merck
  • An efficient synthesis of a dual PPAR alpha/gamma agonist and the formation of a sterically congested alpha-aryloxyisobutyric acid via a Bargellini reaction.
An efficient synthesis of a dual PPAR alpha/gamma agonist and the formation of a sterically congested alpha-aryloxyisobutyric acid via a Bargellini reaction.
The Journal of organic chemistry (2005-10-08)
Raymond J Cvetovich, John Y L Chung, Michael H Kress, Joseph S Amato, Louis Matty, M David Weingarten, Fuh-Rong Tsay, Zhen Li, George Zhou
ABSTRACT
A practical synthesis of benzisoxazole 1 and its conversion to alpha-aryloxyisobutyric acid 2 using 1,1,1-trichloro-2-methyl-2-propanol (chloretone) was developed. Benzisoxazole 1 was formed in high yields by the action of either methanesulfonyl chloride/base upon intermediate oxime 8 or with thionyl chloride/base, which initially forms cyclic sulfite 10. A highly reactive, short-lived intermediate derived from chloretone was detected by ReacIR and its half-life determined to be approximately 5 min. Reaction conditions for the Bargellini reaction were developed that resulted in a 95% yield of 2 from the reaction of highly hindered phenol 1 with chloretone hemihydrate and powdered NaOH in acetone. Thus highly hindered alpha-aryloxyisobutyric acids can be made in a single step in high yield.
Sigma-Aldrich
1,1,1-Trichloro-2-methyl-2-propanol hemihydrate, 98%