Merck
  • Cerebro-hepato-renal syndrome of Zellweger. A report of eight cases with comments upon the incidence, the liver lesion, and a fault in pipecolic acid metabolism.
Cerebro-hepato-renal syndrome of Zellweger. A report of eight cases with comments upon the incidence, the liver lesion, and a fault in pipecolic acid metabolism.
The Journal of pediatrics (1975-03-01)
D M Danks, P Tippett, C Adams, P Campbell
ABSTRACT
Studies of eight patients with this syndrome confirm the characteristic pattern of abnormalities and the autosomal recessive mode of inheritance. The incidence is estimated to be approximately 1 in 100,000 live births. The liver lesion proved to be variable, with features of progressive parenchymal damage rather than a developmental defect of small bile ducts as previously suggested. Elevated levels of pipecolic acid were found in blood and urine and may be related to the basic defect.
Sigma-Aldrich
Pipecolinic acid, 98%