Merck
  • Mapping drug interactions at the covalent topoisomerase II-DNA complex by bisantrene/amsacrine congeners.
Mapping drug interactions at the covalent topoisomerase II-DNA complex by bisantrene/amsacrine congeners.
The Journal of biological chemistry (1998-05-28)
G Capranico, F Guano, S Moro, G Zagotto, C Sissi, B Gatto, F Zunino, E Menta, M Palumbo
ABSTRACT
To identify structural determinants for the sequence-specific recognition of covalent topoisomerase II-DNA complexes by anti-cancer drugs, we investigated a number of bisantrene congeners, including a 10-azabioisoster, bearing one or two 4, 5-dihydro-1H-imidazol-2-yl hydrazone side chains at positions 1, 4, or 9 of the anthracene ring system. The studied bisantrene/amsacrine (m-AMSA) hybrid and bisantrene isomers were able to poison DNA topoisomerase II with an intermediate activity between those of bisantrene and m-AMSA. Moving the side chain from the central to a lateral ring (from C-9 to C-1/C-4) only slightly modified the drug DNA affinity, whereas it dramatically affected local base preferences of poison-stimulated DNA cleavage. In contrast, switching the planar aromatic systems of bisantrene and m-AMSA did not substantially alter the sequence specificity of drug action. A computer-assisted steric and electrostatic alignment analysis of the test compounds was in agreement with the experimental data, since a common pharmacophore was shared by bisantrene, m-AMSA, and 9-substituted analogs, whereas the 1-substituted isomer showed a radically changed pharmacophoric structure. Thus, the relative space occupancy and electron distribution of putative DNA binding (aromatic rings) and enzyme binding (side chains) moieties are fundamental in directing the specific action of topoisomerase II poisons and in determining the poison pharmacophore.
Sigma-Aldrich
Bisantrene dihydrochloride, ≥98% (HPLC)