Merck
  • Comparison of structurally different allosteric modulators of muscarinic receptors by self-organizing neural networks.
Comparison of structurally different allosteric modulators of muscarinic receptors by self-organizing neural networks.
Journal of molecular graphics (1996-08-01)
U Holzgrabe, M Wagener, J Gasteiger
ABSTRACT
Similarities in the molecular structure and surface properties of the allosteric modulators of muscarinic receptors, alcuronium, gallamine, tubocurarine, and the hexamethonium compound W84, a well-known pharmacological tool, are explored. The analysis of the molecular electrostatic potential (MEP) as well as of the shape of the molecular surface is performed by self-organizing neural networks. A distorted sandwich conformation of W84 is suggested to be the active form. The importance of the MEP for binding of these compounds could be established.