Merck
  • Automated on-line dialysis for sample preparation for gas chromatography: determination of benzodiazepines in human plasma.
Automated on-line dialysis for sample preparation for gas chromatography: determination of benzodiazepines in human plasma.
Journal of pharmaceutical and biomedical analysis (1996-06-01)
R Herráez-Hernández, A J Louter, N C van de Merbel, U A Brinkman
ABSTRACT
An on-line dialysis-solid-phase extraction-gas chromatographic (GC) approach has been developed for the determination of drugs in plasma, using some benzodiazepines as model compounds. Clean-up is based on performing the dialysis of 100 microliters samples for 7 min using water as acceptor phase and trapping the diffused analytes on a PLRP-S copolymer precolumn. After drying of the precolumn with nitrogen for 15 min, the analytes are desorbed with ethyl acetate (275 microliters) and injected on-line into the GC system via a loop-type interface. The system provides a very efficient clean-up, and offers the possibility of adding chemical agents which can help to reduce drug-protein binding and, thus, increase sensitivity. To demonstrate the potential of the described approach, the determination of benzodiazepines in plasma at their therapeutical levels is used as an example with flame ionization, thermionic and mass-selective detection.
Product Description
Supelco
N-Desmethylclobazam solution, 100 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®