Merck
  • Effects of nitric oxide synthase inhibitor NG-nitro-L-arginine methyl ester on duodenal alkaline secretory and ulcerogenic responses induced by mepirizole in rats.
Effects of nitric oxide synthase inhibitor NG-nitro-L-arginine methyl ester on duodenal alkaline secretory and ulcerogenic responses induced by mepirizole in rats.
Digestive diseases and sciences (1995-03-01)
K Takeuchi, T Ohuchi, S Okabe
ABSTRACT
The inhibition of nitric oxide (NO) production by NO synthase inhibitors stimulates HCO3- secretion in the rat duodenal mucosa. Therefore, we examined the effects of NG-nitro-L-arginine methyl ester (L-NAME, the NO synthase inhibitor) and nitroprusside (the exogenous NO donor) on the duodenal HCO3- and ulcerogenic responses in anesthetized rats. Animals were administered mepirizole (200 mg/kg, subcutaneously) for induction of duodenal ulcers, and gastric acid and duodenal HCO3- secretions were measured with or without pretreatment with L-NAME (5 mg/kg, intravenously) or nitroprusside (4 mg/kg, intravenously). Mepirizole increased acid secretion, decreased the acid-induced duodenal HCO3- secretion, and induced hemorrhagic lesions in the proximal duodenum. The inhibition of NO production by L-NAME potentiated the acid secretory response, increased the duodenal HCO3- secretion, and prevented the duodenal lesions, and these changes were all antagonized by simultaneous administration of L-arginine (200 mg/kg, intravenously) but not D-arginine. On the other hand, nitroprusside slightly reduced the acid response but further decreased the HCO3- output, resulting in aggravation of duodenal lesions induced by mepirizole. These data suggest that the inhibition of endogenous NO production by the NO synthase inhibitor L-NAME increases duodenal HCO3- secretion and protects the duodenal mucosa against acid injury.
Product Description
Supelco
Mepirizole, analytical standard