Merck
  • Inhibition of brain AChE in brook trout by aminocarb and its toxic metabolites.
Inhibition of brain AChE in brook trout by aminocarb and its toxic metabolites.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes (1985-10-01)
S Y Szeto, K M Sundaram, J Feng
ABSTRACT
In-vitro inhibitions of brain AChE in brook trout, Salvelinus fontinalis (Mitchill), by aminocarb (4-dimethylamino-m-tolyl N-methylcarbamate) and its toxic metabolites, MAA (4-methylamino-m-tolyl N-methylcarbamate), AA(4-amino-m-tolyl N-methylcarbamate), MFA (4-methylformamido-m-tolyl N-methylcarbamate) and FA (4-formamido-m-tolyl N-methylcarbamate) were investigated. The molar concentrations of inhibitors causing 50% inhibition (I50s) were AA (3.62 X 10(-6] less than MAA (7.92 X 10(-6] less than aminocarb (1.01 X 10(-5] less than MFA (4.29 X 10(-5] less than FA (7.11 X 10(-5]. After exposure of fish to various concentrations of aminocarb (25, 250 and 2500 ppb) and MAA (25, 250, 500 and 2500 ppb) at 9 degrees C in dechlorinated tap water for 96 h, inhibitions of brain AChE ranged from 13 to 77%. Mortality occurred only in fish exposed to 500 ppb (22%) and 2500 ppb (100%) of MAA. Enzyme activities recovered to the control levels 12 to 96 h after the fish had been transferred to clean water for clearing; in survivors of the 500-ppb MAA exposure, however, AChE activities decreased again thereafter.
Product Description
Supelco
Aminocarb, PESTANAL®, analytical standard