Merck
Avidin binding of radiolabeled biotin derivatives.
The Journal of biological chemistry (1988-01-05)
R K Garlick, R W Giese
ABSTRACT
Three N-acyl derivatives of biotinylethylenediamine were prepared: I, biotinylamidoethyl-3-(3-[125I]iodo-4-hydroxyphenyl)propionamide; II, biotinylamidoethyl-[3H]acetamide; and III, biotinylamidoethyl-3-(3,5-[125I]diiodo-4-hydroxyphenyl)propionamid e. Each compound was combined with a large excess of avidin, yielding 1:1 molar complexes. Aside from a small fraction of each complex that dissociated more rapidly, the dissociation half-lives of these complexes were: I, 41 days; II, 4.4 days; and III, 148 days. The iodo- (mono or di) hydroxyphenylpropionyl moieties of I and III, therefore, contribute significantly to the binding strength of these compounds toward avidin. We also formed 4:1 complexes of I, II, and III with avidin (compound in excess), each of which exhibited biphasic dissociation, with initial half-lives of 4, 3.2, and 24 days, respectively. Thus, I or especially III potentially can be used as a sensitive tracer in quantitative studies with avidin.
Sigma-Aldrich
N-Biotinyl-ethylenediamine trifluoroacetate salt, ≥96.5% (HPLC)