Merck
Dose-dependent pharmacokinetics of metabolites of dipyrone in saliva.
European journal of drug metabolism and pharmacokinetics (1988-04-01)
E Neddermann, P Rohdewald
ABSTRACT
Metabolites of dipyrone have been determined in the saliva of 18 volunteers following the oral intake of 0.5 g, 1.0 g, 1.5 g, 2.0 g and 2.5 g dipyrone. High concentrations were measured for N-methyl-aminoantipyrine (MAA), the other analgetic active metabolite, 4-aminoantipyrine was found in minor quantities only. N-formyl-antipyrine was not present in the saliva of some volunteers after low doses but could be determined following the intake of doses from 1.5 g - 2.5 g in saliva of all volunteers. N-acetyl-antipyrine appears late in saliva, acetylation rates varied greatly interindividually. Concentrations of the metabolites in saliva mimic the respective non-protein bound fraction in plasma found by other authors.