Merck
  • [Caudal-epidural injection of adjuvants and local anaesthetics in the newborns undergoing surgery].
[Caudal-epidural injection of adjuvants and local anaesthetics in the newborns undergoing surgery].
Klinichna khirurhiia (2013-08-31)
M Iu Kurochkin
ABSTRACT
In 45 newborns, operated on for the inborn failures of gut, the markers of stress and indices of central hemodynamics were studied up. General anaesthesia was applied together with caudal-epidural injection of local anaesthetics in 15 newborns, in 15--the adjuvants--klophelline and promedol--were added to local anaesthetics, in 15--a standard atharalgesia was used. The best and most durable effect was noted while application of anesthesia in combination with adjuvants.
Product Description
Supelco
Fentanyl solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Fentanyl solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Clonidine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®