- Self-inflicted testicular amputation in first lysergic acid diethylamide use.
Self-inflicted testicular amputation in first lysergic acid diethylamide use.
Journal of addiction medicine (2012-12-12)
Christian Blacha, Markus M Schmid, Maximilian Gahr, Roland W Freudenmann, Paul L Plener, Florian Finter, Bernhard J Connemann, Carlos Schönfeldt-Lecuona
PMID23222128
ABSTRACT
Genital self-mutilation is a well-known phenomenon in patients with schizophrenia and has occasionally been described in patients with personality disorders or transsexuality. However, literature just provides few cases of genital self-mutilation related to the use of psychotropic substances. A previously mentally healthy man (age, 32 years) performed manual amputation of both testes after first use of lysergic acid diethylamide in combination with alcohol consumption. Follow-up examination 6 month after the event did not reveal the development of a psychiatric disorder. This report shows that a first and single use of lysergic acid diethylamide in combination with alcohol can cause intoxication with dramatic consequences.
Product Description
Supelco
LSD solution, 25 μg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
LSD solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®