Merck
Actively Q-switched 2.9 μm Ho(3+)Pr(3+)-doped fluoride fiber laser.
Optics letters (2012-06-05)
Tomonori Hu, Darren D Hudson, Stuart D Jackson
ABSTRACT
We report an efficient Q-switched Ho(3+)Pr(3+)-doped fluoride fiber laser, producing a peak power of 77 W, with pulse width of 78 ns. A slope efficiency of 20% with respect to the launched pump power was achieved. A TeO(2) acousto-optic modulator allowed continuous tunability of the pulse repetition frequency from 40 to 300 kHz.
Sigma-Aldrich
Holmium, chips, 99.9% trace metals basis