Merck
  • Role of the brain dopaminergic and opioid system in the regulation of "child's" (maternal bonding) behavior of newborn albino rats.
Role of the brain dopaminergic and opioid system in the regulation of "child's" (maternal bonding) behavior of newborn albino rats.
Bulletin of experimental biology and medicine (2011-01-18)
I S Stovolosov, V A Dubynin, A A Kamensky
ABSTRACT
Administration of D(2) receptor antagonist clebopride in a dose not affecting locomotor activity was followed by a decrease in maternal bonding behavior of 10-day-old and 15-day-old albino rat pups. D(1) receptor antagonist SCH23390 had a stimulatory effect only on the behavior of 10-day-old newborns. Opioid peptide β-casomorphin-7 abolished the effect of clebopride in rat pups of the older age group.