- Catharanthine C16 substituent effects on the biomimetic coupling with vindoline: preparation and evaluation of a key series of vinblastine analogues.
Catharanthine C16 substituent effects on the biomimetic coupling with vindoline: preparation and evaluation of a key series of vinblastine analogues.
Bioorganic & medicinal chemistry letters (2010-10-12)
Annie Tam, Hiroaki Gotoh, William M Robertson, Dale L Boger
PMID20932748
ABSTRACT
The examination of the catharanthine C16 substituent effects on the Fe(III)-promoted biomimetic coupling reaction with vindoline is detailed, confirming the importance of the presence of a C16 electron-withdrawing substituent, and establishing an unanticipated unique role (>10-fold) that the C16 methyl ester plays in the expression of the natural product properties. Thus, replacement of the vinblastine C16' methyl ester with an ethyl ester (10-fold), a cyano group (100-fold), an aldehyde (100-fold), a hydroxymethyl group (1000-fold) or a primary carboxamide (>1000-fold) led to surprisingly large reductions in cytotoxic activity.