Merck
  • [Simultaneous determination of opioid compounds in human urine by UPLC-MS/MS].
[Simultaneous determination of opioid compounds in human urine by UPLC-MS/MS].
Fa yi xue za zhi (2010-08-17)
Chang-Shu Zhan, Bao-Hua Shen, Xian-Yi Zhuo
ABSTRACT
To propose a method for simultaneous determination of codeine(COD), 6-monoacetyl-morphine (6-MAM), morphine (MOR), morphine-3-glucuronide (M3G) and morphine-6-glucuronide (M6G) in human urine by ultra performance liquid chromatography with tandem mass spectrometry (UPLC-MS/MS). After precipitation of protein by acetonitrile, the urine samples, with added the morphine-d3 (MOR-d3) and morphine-3-Glucuronide-d3 (M3G-d3) as internal standards, were pre-treated by Sirocco protein precipitation plate, and then analyzed by UPLC-MS/MS. The limit of detection was 0.2 ng/mL for both COD and MAM, the limit of quantitation was 0.5 ng/mL for both COD and MAM. The limit of detection was 0.5 ng/mL for MOR, M3G and M6G, the limit of quantitation was 1 ng/mL for them. The linear correlation coefficients were not less than 0.9997, both the inter-day and intra-day precisions were less than 10%, the recoveries were in the range of 70.0% to 98.3%, the matrix effects were about 50.5% to 99.0%. This proposed method is simple, rapid and accurate, it could be applied in forensic toxicological analysis.
Product Description
Supelco
Morphine-6-β-D-glucuronide solution, 1.0 mg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-6-β-D-glucuronide solution, 100 μg/mL in methanol: water (1:1), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-3-β-D-glucuronide solution, 100 μg/mL in methanol: water ( 50:50(v/v)), ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Morphine-3-β-D-glucuronide solution, 1 mg/mL in methanol with 0.05% NaOH, ampule of 1 mL, certified reference material, Cerilliant®