Merck
  • Prolyl aminopeptidase from Streptomyces thermoluteus subsp. fuscus strain NBRC14270 and synthesis of proline-containing peptides by its S144C variant.
Prolyl aminopeptidase from Streptomyces thermoluteus subsp. fuscus strain NBRC14270 and synthesis of proline-containing peptides by its S144C variant.
Applied and environmental microbiology (2010-08-03)
Yukihiro Yamamoto, Hirokazu Usuki, Masaki Iwabuchi, Tadashi Hatanaka
ABSTRACT
We specifically examined an exopeptidase, prolyl aminopeptidase (PAP), as a target for synthesis of proline-containing peptides. A PAP from Streptomyces thermoluteus subsp. fuscus NBRC14270 (PAP14270) was obtained using sequence-based screening. From PAP14270, 144Ser was replaced by Cys (scPAP14270) to give aminolysis activity. In contrast to wild-type PAP14270, scPAP14270 produced a polymer of proline benzyl ester and cyclo[Pro-Pro]. The product mass was confirmed using liquid chromatography-mass spectrometry (LC/MS). Several factors affecting the reaction, such as the pH, concentration of the substrate, and reaction time, were measured to determine their effects. Furthermore, a correlation was found between substrate specificity in proline peptide synthesis and the log D value of acyl acceptors in aminolysis catalyzed by scPAP14270. Results showed that dipeptide synthesis proceeded in a weakly acidic environment and that cyclization and polymerization occurred under alkaline conditions. Furthermore, results suggest that almost all amino acid esters whose log D value is greater than 0, except hydroxyproline benzyl ester (Hyp-OBzl), can be recognized as acyl acceptors. These findings support the use of PAPs as a tool for production of physiologically active proline peptides.
Sigma-Aldrich
L-Proline benzyl ester hydrochloride, 98%