Merck
  • A pilot study using nabilone for symptomatic treatment in Huntington's disease.
A pilot study using nabilone for symptomatic treatment in Huntington's disease.
Movement disorders : official journal of the Movement Disorder Society (2009-10-22)
Adrienne Curtis, Ian Mitchell, Smitaa Patel, Natalie Ives, Hugh Rickards
ABSTRACT
Pilot study of nabilone in Huntington's disease (HD). Double-blind, placebo-controlled, cross-over study of nabilone versus placebo. Primary outcome, Unified Huntington's Disease Rating Scale (UHDRS) total motor score. Secondary measures: UHDRS subsections for chorea, cognition and behavior, and neuropsychiatric inventory (NPI). 44 randomized patients received either nabilone (1 or 2 mg) followed by placebo (n = 22), or placebo followed by nabilone (n = 22). Recruiting was straightforward. Nabilone safe and well tolerated, no psychotic episodes. Assessment of either dose of nabilone versus placebo showed a treatment difference of 0.86 (95% CI: -1.8 to 3.52) for total motor score; 1.68 (95% CI: 0.44 to 2.92) for chorea; 3.57 (95% CI: -3.41 to 10.55) for UHDRS cognition; 4.01 (95% CI: -0.11 to 8.13) for UHDRS behavior, and 6.43 (95% CI: 0.2 to 12.66) for the NPI. Larger longer RCT of nabilone in HD is feasible and warranted.
Sigma-Aldrich
Nabilone, solid, ≥98% (HPLC)