Merck
  • Highly enantioselective borane reduction of heteroaryl and heterocyclic ketoxime ethers catalyzed by novel spiroborate ester derived from diphenylvalinol: application to the synthesis of nicotine analogues.
Highly enantioselective borane reduction of heteroaryl and heterocyclic ketoxime ethers catalyzed by novel spiroborate ester derived from diphenylvalinol: application to the synthesis of nicotine analogues.
The Journal of organic chemistry (2008-05-02)
Kun Huang, Francisco G Merced, Margarita Ortiz-Marciales, Héctor J Meléndez, Wildeliz Correa, Melvin De Jesús
ABSTRACT
An asymmetric synthesis for the preparation of nonracemic amines bearing heterocyclic and heteroaromatic rings is described. A variety of important enantiopure thionyl and arylalkyl primary amines were afforded by the borane-mediated enantioselective reduction of O-benzyl ketoximes using 10% of catalyst 10 derived from ( S)-diphenylvalinol and ethylene glycol with excellent enantioselectivity, in up to 99% ee. The optimal condition for the first asymmetric reduction of 3- and 4-pyridyl-derived O-benzyl ketoxime ethers was achieved using 30% of catalytic loading in dioxane at 10 degrees C. ( S)- N-ethylnornicotine ( 3) was also successfully synthesized from the TIPS-protected ( S)-2-amino-2-pyridylethanol in 97% ee.