Merck
Toxicokinetics of aromatic amines in guppy, Poecilia reticulata.
The Science of the total environment (1991-12-01)
W de Wolf, E Yedema, W Seinen, J L Hermens
ABSTRACT
In this study the elimination kinetics of several chlorinated anilines and a tetrachlorobenzene in guppy, Poecilia reticulata, have been determined. The elimination of all the chlorinated anilines in fish showed a bi-phasic pattern, whereas the elimination characteristics of the tetrachlorobenzene are in agreement with a one-compartment model. Elimination of the chlorinated anilines is faster than elimination of the tetrachlorobenzene. The small differences in octanol-water partition coefficient of these compounds can only in part describe the differences in elimination rate. The observed differences might be attributed to differing biotransformation rates.
Product Description
Supelco
Pentachloroaniline, PESTANAL®, analytical standard