Merck
  • Biosensor-based kinetic characterization of the interaction between HIV-1 reverse transcriptase and non-nucleoside inhibitors.
Biosensor-based kinetic characterization of the interaction between HIV-1 reverse transcriptase and non-nucleoside inhibitors.
Journal of medicinal chemistry (2006-04-14)
Matthis Geitmann, Torsten Unge, U Helena Danielson
ABSTRACT
Details of the interaction between HIV-1 reverse transcriptase and non-nucleoside inhibitors (NNRTIs) have been elucidated using a biosensor-based approach. This initial study was performed with HIV-1 reverse transcriptase mutant K103N, the phenethylthioazolylthiourea compound (PETT) MIV-150, and the three NNRTIs licensed for clinical use: nevirapine, delavirdine, and efavirenz. Mathematical evaluation of the experimental data with several interaction models revealed that the four inhibitors interacted with HIV-1 RT with varying degrees of complexity. The simplest adequate model accounted for two different conformations of the free enzyme, of which only one can bind the inhibitor, consistent with a previously hypothesized population-shift model including a preformation of the NNRTI binding site. In addition, a heterogeneous binding was observed for delavirdine, efavirenz, and MIV-150, indicating that two noncompetitive and kinetically distinct enzyme-inhibitor complexes could be formed. Furthermore, for these compounds, there were indications for ligand-induced conformational changes.
Sigma-Aldrich
Delavirdine mesylate, ≥98% (HPLC)