Merck
  • Glomerular tip lesion associated with nonsteroidal anti-inflammatory drug-induced nephrotic syndrome.
Glomerular tip lesion associated with nonsteroidal anti-inflammatory drug-induced nephrotic syndrome.
American journal of kidney diseases : the official journal of the National Kidney Foundation (2005-09-27)
Inderpreet Sekhon, Sandeep Munjal, Byron Croker, Richard J Johnson, A Ahsan Ejaz
ABSTRACT
Glomerular tip lesion and its relation to different glomerular diseases is a subject of controversy. The therapeutic and prognostic clinical implications of glomerular tip lesions are ambiguous. We present a case of glomerular tip lesion associated with nonsteroidal anti-inflammatory drug-induced nephrotic syndrome that further complicates this issue. To our knowledge, this is the first case report of glomerular tip lesion associated with nonsteroidal anti-inflammatory drug-induced nephrotic syndrome.
Product Description
Supelco
Butalbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.