Merck
  • Response of growth, photosynthesis and photoinhibition of the edible cyanobacterium Nostoc sphaeroides colonies to thiobencarb herbicide.
Response of growth, photosynthesis and photoinhibition of the edible cyanobacterium Nostoc sphaeroides colonies to thiobencarb herbicide.
Chemosphere (2005-03-25)
Jianrong Xia
ABSTRACT
Effects of thiobencarb on growth, photosynthesis and photoinhibition of Nostoc sphaeroides colonies were examined. Thiobencarb concentrations higher than 2 mg/l led to a significant decrease in phycoerythrin, phycocyanin and allophycocyanin content, but no significant effect on chlorophyll a (chl a) content. Biomass yield, protein content and photosynthetic rate were reduced only in the highest thiobencarb concentration (10 mg/l). Colonies exposed to high thiobencarb dose (10 mg/l) were more sensitive to high light than those exposed to the control and 4 mg/l thiobencarb. Dim light induced recovery from photoinhibition to more great extent in colonies grown in the control and 4 mg/l thiobencarb than 10 mg/l thiobencarb. Darkness resulted in a insignificant recovery regardless of thiobencarb treatments.
Product Description
Supelco
Thiobencarb, PESTANAL®, analytical standard