Merck
  • Overcoming blood shortages: red blood cell and platelet substitutes and membrane modifications.
Overcoming blood shortages: red blood cell and platelet substitutes and membrane modifications.
Current hematology reports (2004-02-18)
Richard J Davey
ABSTRACT
Blood shortages are increasingly common as the donor base declines and extensive restrictions on blood donation disqualify many donors. Red blood cell (RBC) and platelet substitutes have long been anticipated as alternatives to standard transfusions. However, difficulties in manufacturing, efficacy, and safety have slowed the development of these products. New understanding of the relationship between blood viscosity, oxygen transport, and vasoactivity have led to more effective RBC substitutes, several of which are in advanced clinical trials. In addition, creative approaches to RBC membrane modification, such as the enzymatic cleavage of ABH glycoproteins, may lead to a universal RBC. Advances in the understanding of platelet membrane behavior at low temperatures may lead to extended platelet storage at refrigerator temperatures. Standard transfusions of human RBCs and platelets will not be replaced soon. However, these new products will be a useful alternative for selected clinical applications and will lessen our dependence on our marginally adequate blood supply.