Merck
  • Effects of dietary ingredients on function and expression of P-glycoprotein in human intestinal epithelial cells.
Effects of dietary ingredients on function and expression of P-glycoprotein in human intestinal epithelial cells.
Biological & pharmaceutical bulletin (2010-02-02)
Takashi Okura, Michiko Ibe, Keizo Umegaki, Kazumasa Shinozuka, Shizuo Yamada
ABSTRACT
The present study was conducted to investigate the functional and transcriptional modulation of P-glycoprotein (MDR-1) by several dietary ingredients (piperine, capsaicin, daidzein, genistein, sesamin, curcumin, taurine) in vinblastine-resistant colon carcinoma LS-180 cells (LS-180V cells). The amount of rhodamine 123 accumulated in LS-180V cells was significantly increased by capsaicin, piperine and sesamin, whereas it was significantly reduced by daidzein and genistein which stimulated the efflux of rhodamine 123. These results suggest that the P-glycoprotein-mediated efflux is inhibited by piperine, capsaicin and sesamin and stimulated by daidzein and genistein. The concurrent addition of piperine and capsaicin seemed to inhibit synergistically the P-glycoprotein-mediated efflux. Pretreatment with sesamin for 48 h caused a significant increase in MDR1 mRNA expression without a significant effect on the expression of P-glycoprotein or accumulation of rhodamine 123. Similar pretreatment with other ingredients had little effect on the expression of MDR1 mRNA or P-glycoprotein, suggesting that they do not cause transcriptional modulation of P-glycoprotein. Piperine, genistein and curcumin have been suggested to stimulate P-glycoprotein-mediated efflux without increasing P-glycoprotein expression. In LS-180V cells, significant increases in mRNA levels of multi-drug resistance associated protein 1 (MRP1) or MRP3 were observed on pretreatment with capsaicin, daidzein, piperine and sesamin. In conclusion, our results suggest that P-glycoprotein-mediated efflux is significantly affected by dietary ingredients. Also, capsaicin, daidzein, piperine and sesamin increased significantly the mRNA expression of MRP1 or MRP3. Thus, the present study provides further evidence that repeated exposure to dietary ingredients can cause drug-food interactions by affecting the function and mRNA expression of intestinal transporters such as P-glycoprotein.
Product Description
Supelco
Daidzin, analytical standard
Sigma-Aldrich
Daidzin, ≥95.0% (HPLC)