Merck
  • [Potentiometric electrodes for determining choline, butyrylcholine and cholinesterase inhibitors].
[Potentiometric electrodes for determining choline, butyrylcholine and cholinesterase inhibitors].
Prikladnaia biokhimiia i mikrobiologiia (1998-06-30)
A L Gindilis, I N Kurochkin
ABSTRACT
Potentiometric choline electrodes were developed on the basis of the mediator-free bioelectrocatalysis. The electrodes made of a composite carbon-polymer material contain choline oxidase and peroxidase coimmobilized on the surface of the electrode. The rate of the potential increase was shown to be proportional to the choline concentration within a broad range of variation. Coupling of choline-sensitive electrodes with butyrylcholinesterase makes possible both the direct detection of butyrylcholine and analysis of butyrylcholinesterase inhibitors.
Sigma-Aldrich
Butyrylcholine chloride, ≥98%