Merck
  • Large-scale synthesis of the catechol metabolites of diethylstilbestrol and hexestrol.
Large-scale synthesis of the catechol metabolites of diethylstilbestrol and hexestrol.
Chemical research in toxicology (1998-06-20)
S T Jan, E G Rogan, E L Cavalieri
ABSTRACT
Diethylstilbestrol (DES) and hexestrol (HES) are carcinogenic synthetic estrogens. The major metabolites of these compounds are their catechol derivatives, 3'-OH-DES and 3'-OH-HES. Oxidation of these metabolites leads to the electrophilic quinones, which are presumably involved in the tumor-initiating process. A synthetic route based on the McMurry coupling reaction was developed for the synthesis of 3'-OH-DES. Using commercially inexpensive starting materials, this compound was synthesized in four steps, and the cis and trans isomers were separated and identified. Following the same synthetic route, 3'-OH-HES was synthesized in five steps.