MilliporeSigma
  • Evaluation of skin irritation and sensitization of two diol solutions used as experimental dentin primers in humans and guinea pigs.
Evaluation of skin irritation and sensitization of two diol solutions used as experimental dentin primers in humans and guinea pigs.
Dental materials journal (1996-12-01)
A Kurihara, A Manabe, K Katsuno, K Itoh, H Hisamitsu, S Wakumoto, T Yoshida
ABSTRACT
The purpose of the present study was to evaluate the safety of ethylene glyco (EG) and 1,6-hexanediol (HD) solutions as experimental dentin primers when subjected to the guinea pig maximization test (GPMT), primary irritation test, cumulative skin irritation test and human patch test. No primary and cumulative skin irritation resulting from the use of 62.5% EG or 45% HD solutions was observed. In the case of GPMT, the animals sensitized with 2-hydroxyethyl methacrylate (2-HEMA) responded to 100% HD. 62.5% EG and 45% HD as dentin primers were safer than 2-HEMA such as a methacrylic primer.
Sigma-Aldrich
1,6-Hexanediol, 97%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,6-Hexanediol, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
1,6-Hexanediol solution, BioUltra, ~6 M in H2O