Merck
  • Evaluation of anti-calculus efficacy. An SEM method of evaluating the effectiveness of pyrophosphate dentifrice on calculus formation.
Evaluation of anti-calculus efficacy. An SEM method of evaluating the effectiveness of pyrophosphate dentifrice on calculus formation.
Journal of clinical periodontology (1993-02-01)
P Gaengler, A Kurbad, W Weinert
ABSTRACT
A double-blind, cross-over study was carried out to assess the anti-calculus effectiveness of a pyrophosphate containing cleaning gel. 2 groups of 30 heavy calculus formers were followed up comparing conventional clinical scoring systems (Volpe-Manhold-Index and Marginal-Line-Calculus-Index) and micromorphologic SEM criteria of calculus formation and inhibition. The results indicated that after a 3-month period, there was a significant reduction of calculus formation compared to a placebo gel. The proposed SEM calculus index (SEMCI) objectively reflects the reduction of supragingival calculus formation.
Sigma-Aldrich
Potassium pyrophosphate, 97%