Merck
  • Sample preparation of carbadox, furazolidone, nitrofurazone, and ethopabate in medicated feeds for high pressure liquid chromatography.
Sample preparation of carbadox, furazolidone, nitrofurazone, and ethopabate in medicated feeds for high pressure liquid chromatography.
Journal - Association of Official Analytical Chemists (1980-09-01)
V A Thorpe
ABSTRACT
Medicated feeds (pelleted or mash) containing guarantees of carbadox, furazolidone, nitrofurazone, and ethopabate are pretreated with water, extracted with 95% dimethylformamide overnight at room temperature, cleaned up on a column of alumina, and injected into a high pressure liquid chromatograph for quantitative measurement. Carbadox, nitrofurazone, and furazolidone can be separated; chromatograms show excellent baseline resolution, and results are in good agreement with colorimetric methods. The same extraction and cleanup can be used to improve colorimetric methods for furazolidone and nitrofurazone.
Product Description
Supelco
Ethopabate, VETRANAL®, analytical standard