Merck
  • Bacterial metabolism of alpha-pinene: pathway from alpha-pinene oxide to acyclic metabolites in Nocardia sp. strain P18.3.
Bacterial metabolism of alpha-pinene: pathway from alpha-pinene oxide to acyclic metabolites in Nocardia sp. strain P18.3.
Journal of bacteriology (1987-11-01)
E T Griffiths, S M Bociek, P C Harries, R Jeffcoat, D J Sissons, P W Trudgill
ABSTRACT
Over 20 gram-positive bacteria were isolated by elective culture with (+/-)-alpha-pinene as the sole carbon source. One of these strains, Nocardia sp. strain P18.3, was selected for detailed study. alpha-Pinene-grown cells oxidized, without lag, alpha-pinene, alpha-pinene oxide (epoxide), and the cis and trans isomers of 2-methyl-5-isopropylhexa-2,5-dienal. No other tested terpene was oxidized at a significant rate. alpha-Pinene was not metabolized by cell extracts in the presence or absence of NADH or NADPH. Cell extracts catalyzed a rapid decyclization of alpha-pinene oxide, in the absence of added cofactors, with the formation of cis-2-methyl-5-isopropylhexa-2,5-dienal. Further oxidation of the aldehyde to the corresponding acid occurred in the presence of NAD. Both activities were induced by growth with alpha-pinene. A rapid, nonenzymic transformation of the cis aldehyde into the trans isomer occurred in glycine buffer. The trans isomer was also a substrate for the NAD-linked aldehyde dehydrogenase. The distribution of the alpha-pinene oxide lyase in alpha-pinene-utilizing Pseudomonas spp. was also investigated and was compatible with the two alternative ring-cleavage sequences that have been proposed on the basis of accumulated metabolites.
Sigma-Aldrich
α-Pinene oxide, 97%