Merck
Sequence-controlled polymers.
Science (New York, N.Y.) (2013-08-10)
Jean-François Lutz, Makoto Ouchi, David R Liu, Mitsuo Sawamoto
ABSTRACT
Sequence-controlled polymers are macromolecules in which monomer units of different chemical nature are arranged in an ordered fashion. The most prominent examples are biological and have been studied and used primarily by molecular biologists and biochemists. However, recent progress in protein- and DNA-based nanotechnologies has shown the relevance of sequence-controlled polymers to nonbiological applications, including data storage, nanoelectronics, and catalysis. In addition, synthetic polymer chemistry has provided interesting routes for preparing nonnatural sequence-controlled polymers. Although these synthetic macromolecules do not yet compare in functional scope with their natural counterparts, they open up opportunities for controlling the structure, self-assembly, and macroscopic properties of polymer materials.
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
Taq DNA Polymerase from Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
DNA Polymerase I from Escherichia coli lysogenic for NM 964, buffered aqueous glycerol solution
Sigma-Aldrich
DNA Polymerase I, Klenow Fragment from Escherichia coli, buffered aqueous glycerol solution