Merck
  • Nonsurgical treatment of aggressive periodontitis with photodynamic therapy or systemic antibiotics. Three-month results of a randomized, prospective, controlled clinical study.
Nonsurgical treatment of aggressive periodontitis with photodynamic therapy or systemic antibiotics. Three-month results of a randomized, prospective, controlled clinical study.
Schweizer Monatsschrift fur Zahnmedizin = Revue mensuelle suisse d'odonto-stomatologie = Rivista mensile svizzera di odontologia e stomatologia (2013-07-10)
Nicole B Arweiler, Malgorzata Pietruska, Ania Skurska, Ewa Dolińska, Jan K Pietruski, Maximilian Bläs, Thorsten M Auschill, Anton Sculean
ABSTRACT
The aim of this randomized, controlled clinical study was to compare the short-term effects of nonsurgical periodontal therapy with the additional administration of systemic antibiotics (AB) and the same therapy with additional photodynamic therapy (PDT) in the treatment of patients with aggressive periodontitis (AP). Thirty-six patients with AP received full-mouth nonsurgical periodontal treatment (SRP) and were then randomly divided into two groups of 18 subjects each. Group AB received amoxicillin and metronidazole three times a day for 7 days. Group PDT received two applications of PDT on the day of SRP as well as at follow-up after 7 days. The following clinical parameters were measured at baseline and 3 months after therapy: plaque index (PLI), bleeding on probing (BOP), probing depth (PD), gingival recession (GR), and clinical attachment level (CAL). After 3 months, PD was significantly reduced in both groups (from 5.0±0.8 mm to 3.2±0.4 mm with AB, and 5.1±0.5 mm to 4.0±0.8 mm with PDT; both p<0.001), while AB revealed significantly lower values compared to PDT (p = 0.001). In both groups, GR was not significantly changed. CAL was significantly reduced in both groups (PDT: 5.7±0.8 mm to 4.7±1.1 mm; p=0.011; AB: 5.5±1.1 mm to 3.9±1.0 mm; p<0.001) and differed significantly between the groups (p=0.025). The number of residual pockets (PD ≥4 mm) and positive BOP was reduced by AB from 961 to 377, and by PDT from 628 to 394. Pockets with PD ≥7 mm were reduced by AB from 141 to 7, and by PDT from 137 to 61. After 3 months, both treatments led to statistically significant clinical improvements. The systemic administration of antibiotics, however, resulted in significantly higher reduction of PD and a lower number of deep pockets compared to PDT.
Product Description
Supelco
Metronidazole, VETRANAL®, analytical standard
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Supelco
Amoxicillin trihydrate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Amoxicillin trihydrate, VETRANAL®, analytical standard