Merck
Cellulose contents of some abundant Indian seaweed species.
Natural product communications (2013-06-07)
Arup K Siddhanta, Sanjay Kumar, Gaurav K Mehta, Mahesh U Chhatbar, Mihir D Oza, Naresh D Sanandiya, Dharmesh R Chejara, Chirag B Godiya, Stalin Kondaveeti
ABSTRACT
Crude cellulose as well as alpha- and beta-celluloses were estimated in thirty-four seaweed species of fifteen orders of Chlorophyta, Phaeophyta and Rhodophyta of Indian waters. The greatest yields of crude cellulose and a-cellulose were obtained from Chaetomorpha aerea (approx. 20.0% and 18.5%, respectively), and of beta-cellulose (approx. 3.1%) from Caulerpa imbricata. The lowest crude cellulose, and alpha-and beta-contents were recorded for the calcareous red alga Liagora indica (approx. 0.90%, 0.70% and 0.10%, respectively). There was little variation in cellulose content among the brown algae, while wide variations in the yields were found in the green and red algae. The present work contributes to the repertoire of 67 Indian seaweed species studied to now for their cellulose contents in our laboratory. The combined studies highlight that Chaetomorpha aerea, Acrosiphonia orientalis, Caulerpa taxifolia, Sargassum tenerrimum, Hydroclathrus clathratus and Gelidiella acerosa possess relatively high (> 10%) cellulose contents, which could be of potential utility.
Product Description
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, acid washed, powder, for column chromatography
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Sigma-Aldrich
Cellulose, fibers, (medium)
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
α-Cellulose, powder
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm