Merck
  • Inhibition of RNA helicase Brr2 by the C-terminal tail of the spliceosomal protein Prp8.
Inhibition of RNA helicase Brr2 by the C-terminal tail of the spliceosomal protein Prp8.
Science (New York, N.Y.) (2013-05-25)
Sina Mozaffari-Jovin, Traudy Wandersleben, Karine F Santos, Cindy L Will, Reinhard Lührmann, Markus C Wahl
ABSTRACT
The Ski2-like RNA helicase Brr2 is a core component of the spliceosome that must be tightly regulated to ensure correct timing of spliceosome activation. Little is known about mechanisms of regulation of Ski2-like helicases by protein cofactors. Here we show by crystal structure and biochemical analyses that the Prp8 protein, a major regulator of the spliceosome, can insert its C-terminal tail into Brr2's RNA-binding tunnel, thereby intermittently blocking Brr2's RNA-binding, adenosine triphosphatase, and U4/U6 unwinding activities. Inefficient Brr2 repression is the only recognizable phenotype associated with certain retinitis pigmentosa-linked Prp8 mutations that map to its C-terminal tail. Our data show how a Ski2-like RNA helicase can be reversibly inhibited by a protein cofactor that directly competes with RNA substrate binding.
Sigma-Aldrich
Ribonucleic acid from torula yeast, Type VI
Sigma-Aldrich
Ribonucleic acid from torula yeast, core, Type II-C
Sigma-Aldrich
Ribonucleic acid diethylaminoethanol salt, Type IX
Sigma-Aldrich
Ribonucleic acid from baker′s yeast (S. cerevisiae)