Merck
  • Improved quantification of HIV-1-infected CD4+ T cells using an optimised method of intracellular HIV-1 gag p24 antigen detection.
Improved quantification of HIV-1-infected CD4+ T cells using an optimised method of intracellular HIV-1 gag p24 antigen detection.
Journal of immunological methods (2013-03-19)
Hongbing Yang, Elisabeth Yorke, Gemma Hancock, Genevieve Clutton, Nellia Sande, Brian Angus, Redmond Smyth, Johnson Mak, Lucy Dorrell
ABSTRACT
The capacity of CD8+ T cells to inhibit HIV-1 replication in vitro strongly correlates with virus control in vivo. Post-hoc evaluations of HIV-1 vaccine candidates suggest that this immunological parameter is a promising benchmark of vaccine efficacy. Large-scale analysis of CD8+ T cell antiviral activity requires a rapid, robust and economical assay for accurate quantification of HIV-1 infection in primary CD4+ T cells. Detection of intracellular HIV-1 p24 antigen (p24 Ag) by flow cytometry is one such method but it is thought to be less sensitive and quantitative than p24 Ag ELISA. We report that fixation and permeabilisation of HIV-infected cells using paraformaldehyde/50% methanol/Nonidet P-40 instead of a conventional paraformaldehyde/saponin-based protocol improved their detection across multiplicities of infection (MOI) ranging from 10(-2) to 8×10(-5), and by nearly two-fold (p<0.001) at the optimal MOI tested (10(-2)). The frequency of infected cells was strongly correlated with p24 Ag release during culture, thus validating its use as a measure of productive infection. We were also able to quantify infection with a panel of HIV-1 isolates representing the major clades. The protocol described here is rapid and cost-effective compared with ELISA and thus could be a useful component of immune monitoring of HIV-1 vaccines and interventions to reduce viral reservoirs.
Sigma-Aldrich
Paraformaldehyde, meets analytical specification of DAC, 95.0-100.5%
Sigma-Aldrich
Triton X-114
Sigma-Aldrich
IGEPAL® CA-720, average Mn ~735
Sigma-Aldrich
Paraformaldehyde, powder, 95%
Sigma-Aldrich
Paraformaldehyde, prilled, 95%
Sigma-Aldrich
Paraformaldehyde, reagent grade, crystalline
Sigma-Aldrich
Triton X-165 solution, Non-ionic, CMC-0.43 mM, suitable for electrophoresis, cloud point >100 °C
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology
Sigma-Aldrich
Triton X-114, laboratory grade
Sigma-Aldrich
IGEPAL® CA-630, viscous liquid
Supelco
IGEPAL® CA-630
Sigma-Aldrich
Triton X-405 solution, 70% in H2O