Merck
  • Gene expression profiling in the ovary of Queen conch (Strombus gigas) exposed to environments with high tributyltin in the British Virgin Islands.
Gene expression profiling in the ovary of Queen conch (Strombus gigas) exposed to environments with high tributyltin in the British Virgin Islands.
The Science of the total environment (2013-02-16)
Cassander P Titley-O'Neal, Daniel J Spade, Yanping Zhang, Rosalinda Kan, Christopher J Martyniuk, Nancy D Denslow, Bruce A MacDonald
ABSTRACT
Queen conch (Strombus gigas) are listed in CITES Appendix II. Populations may be declining due to anthropogenic inputs that include pollutants from boating activity. In the British Virgin Islands (BVI), some conch exhibit imposex, a condition in which male external genitalia are present in female conch. Previous studies suggest that tributyltin (TBT), an antifouling chemical used in boat paint, is correlated to increased incidence of imposex although the mechanisms leading to imposex are not known. The present study utilized a Queen conch microarray to measure the response of the ovarian transcriptome in conch inhabiting polluted environments with high TBT levels in the BVI. The polluted sites, Road Harbour (RH) and Trellis Bay (TB), are areas with high boating activity while the reference sites, Guana Island (GI) and Anegada (AN), are areas with low boating activity. There were 754 and 898 probes differentially expressed in the ovary of conch collected at RH and TB respectively compared to conch collected at GI. Of the genes that were differentially expressed at both sites, >10% were shared suggesting that these sites have additional environmental factors influencing gene expression patterns. Functional enrichment analysis showed that the biological processes of cell proliferation, translation, and oxidative stress were over-represented in the polluted sites. Gene set enrichment analysis revealed that transcripts involved in the biological processes of general metabolism, immune, lipid metabolism, and stress were affected in conch from polluted environments. Interestingly, altered stress genes appeared to be more prevalent in conch collected from RH than TB, corresponding to the higher TBT load at RH compared to TB. Our study shows that stress pathways are affected in conch ovary in environments that experience heavy boating activity in the BVIs, although we are unable to directly link changes at the transcriptomics level to high TBT levels.
Sigma-Aldrich
Tributyltin hydride, contains 0.05% BHT as stabilizer, 97%